Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202543.1 | NP_001189472.1 | 24 | 10111 | ||
NM_181552.3 | NP_853530.2 | 24 | 9989 | |||
XM_005250150.2 | XP_005250207.1 | 24 | 10507 | |||
XM_006715854.2 | XP_006715917.1 | 23 | 10444 | |||
XM_011515823.2 | XP_011514125.1 | 24 | 10202 | |||
XM_011515825.1 | XP_011514127.1 | 24 | 9770 | |||
XM_017011760.1 | XP_016867249.1 | 23 | 10004 | |||
GenBank mRNA | AK056307.1 | - | 1 | 1496 |