Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020926.1 | - | 1 | 657 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA571409.1 | - | 1 | 14 | Chr. 4 - 22397865 - 22398680 [-] on Build GRCh38 |