Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300883.2 | NP_001287812.1 | 6 | 7201 | ||
NM_001318055.1 | NP_001304984.1 | 7 | 6937 | |||
NM_001318056.1 | NP_001304985.1 | 7 | 6981 | |||
NM_024106.2 | NP_077011.1 | 8 | 7064 | |||
GenBank mRNA | AK024250.1 | - | 2 | 2250 |