Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300883.2 | NP_001287812.1 | 6 | 6308 | ||
NM_001318055.1 | NP_001304984.1 | 7 | 6044 | |||
NM_001318056.1 | NP_001304985.1 | 7 | 6088 | |||
NM_024106.2 | NP_077011.1 | 8 | 6171 | |||
GenBank mRNA | AK024250.1 | - | 2 | 1356 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU959283.1 | - | 34 | Chr. - - [-] on Build |