Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129578.1 | - | 1 | 89 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BJ992915.1 | - | 1 | 170 | Chr. 1 - 31910870 - 31911361 [-] on Build GRCh38 |