Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320470.1 | NP_001307399.1 | 24 | 2717 | ||
XM_005257472.1 | XP_005257529.1 | 25 | 2713 | |||
XM_011524939.1 | XP_011523241.1 | 28 | 3082 | |||
XM_011524940.1 | XP_011523242.1 | 27 | 3037 | |||
XM_011524941.1 | XP_011523243.1 | 26 | 2947 | |||
XM_011524942.2 | XP_011523244.1 | 26 | 2803 | |||
XM_011524943.2 | XP_011523245.1 | 25 | 2758 | |||
XM_017024783.1 | XP_016880272.1 | 28 | 3082 | |||
XM_017024786.1 | XP_016880275.1 | 27 | 2992 | |||
XM_017024787.1 | XP_016880276.1 | 27 | 2944 | |||
XM_017024790.1 | XP_016880279.1 | 25 | 2713 | |||
XM_017024791.1 | XP_016880280.1 | 24 | 2668 | |||
GenBank mRNA | AJ511332.1 | - | 1 | 12 | ||
AK000135.1 | - | 10 | 1195 | |||
AK125409.1 | - | 1 | 2692 | |||
AK225757.1 | - | 24 | 2676 | |||
AL831895.1 | - | 11 | 1233 |