Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008528.2 | NP_001008528.1 | 4 | 4437 | ||
XM_005257382.2 | XP_005257439.1 | 3 | 4523 | |||
XM_011524830.1 | XP_011523132.1 | 4 | 4323 | |||
GenBank mRNA | AK097108.1 | - | 1 | 2079 |