Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268009.2 | XP_005268066.1 | 11 | 12985 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL833129.1 | - | 1 | 2338 | Chr. 14 - 60140153 - 60143393 [+] on Build GRCh38 |