Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520069.2 | XP_011518371.1 | 7 | 4288 | ||
GenBank mRNA | CR627308.1 | - | 1 | 2567 |