Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013438.2 | NP_001013456.1 | 16 | 8156 | ||
NM_001013439.2 | NP_001013457.1 | 18 | 8592 | |||
NM_005087.3 | NP_005078.2 | 17 | 8248 | |||
GenBank mRNA | AF075028.1 | - | 1 | 243 |