Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519457.1 | XP_011517759.1 | 1 | 108 | ||
XM_017016191.1 | XP_016871680.1 | 1 | 108 | |||
GenBank mRNA | AK022980.1 | - | 1 | 715 |