Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 7 | 2481 | ||
NM_194259.2 | NP_919235.1 | 8 | 2738 | |||
NM_194260.2 | NP_919236.1 | 7 | 2601 | |||
NM_194261.2 | NP_919237.1 | 7 | 2453 | |||
XM_017023640.1 | XP_016879129.1 | 7 | 2878 | |||
GenBank mRNA | BC064492.1 | - | 1 | 1292 |