Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110609.1 | 6 | 457 | |||
GenBank mRNA | BC038533.1 | - | 6 | 457 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI830547.1 | - | 457 | Chr. - - [-] on Build |