Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103184.3 | NP_001096654.1 | 17 | 11854 | ||
NM_001277313.1 | NP_001264242.1 | 21 | 12879 | |||
XM_011521504.2 | XP_011519806.1 | 21 | 12702 | |||
XM_011521506.2 | XP_011519808.1 | 19 | 12616 | |||
XM_011521509.2 | XP_011519811.1 | 20 | 10910 | |||
XM_011521511.2 | XP_011519813.1 | 18 | 12381 | |||
XM_017022130.1 | XP_016877619.1 | 18 | 13073 | |||
XM_017022132.1 | XP_016877621.1 | 18 | 12827 | |||
XM_017022133.1 | XP_016877622.1 | 20 | 12732 | |||
XM_017022134.1 | XP_016877623.1 | 18 | 11283 | |||
XM_017022135.1 | XP_016877624.1 | 18 | 12552 | |||
GenBank mRNA | BC029107.1 | - | 1 | 1174 |