Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006218.3 | NP_006209.2 | 21 | 4897 | ||
GenBank mRNA | AK021510.1 | - | 1 | 108 |