Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712008.3 | XP_006712071.1 | 6 | 1267 | ||
XM_006712009.3 | XP_006712072.1 | 6 | 1264 | |||
XM_006712010.3 | XP_006712073.1 | 5 | 1132 | |||
XM_006712011.3 | XP_006712074.1 | 5 | 1129 | |||
XM_006712012.3 | XP_006712075.1 | 5 | 1129 | |||
XM_006712013.3 | XP_006712076.1 | 4 | 994 | |||
XM_006712014.3 | XP_006712077.1 | 3 | 877 | |||
GenBank mRNA | AB209518.1 | - | 1 | 1201 |