Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102546226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105021.1 | 6 | 1036 | |||
GenBank mRNA | BC043237.1 | - | 6 | 1039 |