Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038373.1 | 4 | 1878 | |||
NR_134458.1 | 3 | 2058 | ||||
GenBank mRNA | BC043233.1 | - | 4 | 1886 |