Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY236490.1 | - | 1 | 57 | ||
AY263163.1 | - | 1 | 20 | |||
BC132679.1 | - | 1 | 15 | 40146701 | ||
BC143410.1 | - | 1 | 15 | 9051917 | ||
BC157874.1 | - | 1 | 27 | 9057092 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX640632.1 | - | 114 | Chr. - - [-] on Build |