Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152485.3 | NP_689698.1 | 2 | 3854 | ||
GenBank mRNA | AK022798.1 | - | 1 | 745 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM723893.1 | - | 1 | 605 | Chr. 1 - 209780165 - 209780813 [-] on Build GRCh38 |