Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715042.2 | XP_006715105.1 | 1 | 673 | ||
GenBank mRNA | AK127022.1 | - | 1 | 673 |