Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104189.1 | 4 | 2194 | |||
GenBank mRNA | AK127439.1 | - | 4 | 2194 |