Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512840.2 | XP_011511142.1 | 8 | 3529 | ||
GenBank mRNA | BX647284.1 | - | 1 | 4149 |