Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190796.2 | NP_001177725.1 | 3 | 3704 | ||
NM_001291999.1 | NP_001278928.1 | 4 | 3974 | |||
NM_006153.5 | NP_006144.1 | 4 | 3940 | |||
GenBank mRNA | AK095534.1 | - | 1 | 1850 |