Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF446131.1 | - | 1 | 73 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE297602.1 | - | 1 | 20 | Chr. 12 - 117331433 - 117452170 [-] on Build GRCh38 |