Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138966.3 | NP_620416.1 | 11 | 4277 | ||
XM_005266774.4 | XP_005266831.1 | 11 | 3796 | |||
XM_017026020.1 | XP_016881509.1 | 11 | 3884 | |||
GenBank mRNA | AK123319.1 | - | 1 | 174 |