Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257291.1 | NP_001244220.1 | 17 | 7917 | ||
NM_032591.2 | NP_115980.1 | 17 | 7914 | |||
XM_005272681.4 | XP_005272738.1 | 18 | 7370 | |||
XM_005272682.4 | XP_005272739.1 | 18 | 7354 | |||
XM_011543990.2 | XP_011542292.1 | 18 | 3270 | |||
XM_017029905.1 | XP_016885394.1 | 18 | 3254 | |||
XM_017029906.1 | XP_016885395.1 | 17 | 7310 | |||
GenBank mRNA | AK096921.1 | - | 1 | 996 | ||
BC090942.1 | - | 1 | 1850 |