Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273032.3 | XP_005273089.1 | 6 | 7514 | ||
XM_011544041.2 | XP_011542343.1 | 7 | 7613 | |||
XM_011544043.2 | XP_011542345.1 | 9 | 7946 | |||
XM_011544044.2 | XP_011542346.1 | 8 | 7717 | |||
XM_011544045.2 | XP_011542347.1 | 8 | 7842 | |||
XM_011544046.2 | XP_011542348.1 | 6 | 7550 | |||
XM_011544047.2 | XP_011542349.1 | 6 | 7486 | |||
XM_017000152.1 | XP_016855641.1 | 8 | 10539 | |||
XM_017000153.1 | XP_016855642.1 | 7 | 7617 | |||
XM_017000154.1 | XP_016855643.1 | 6 | 7311 | |||
XM_017000155.1 | XP_016855644.1 | 7 | 7654 | |||
XM_017000156.1 | XP_016855645.1 | 7 | 7618 | |||
XM_017000157.1 | XP_016855646.1 | 5 | 7219 | |||
XM_017000158.1 | XP_016855647.1 | 5 | 7451 | |||
XM_017000159.1 | XP_016855648.1 | 5 | 7423 | |||
XM_017000160.1 | XP_016855649.1 | 4 | 8273 | |||
XM_017000161.1 | XP_016855650.1 | 6 | 7555 | |||
XM_017000162.1 | XP_016855651.1 | 5 | 7387 | |||
XM_017000163.1 | XP_016855652.1 | 4 | 11249 | |||
GenBank mRNA | AK095715.1 | - | 1 | 2219 |