Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273032.3 | XP_005273089.1 | 6 | 6681 | ||
XM_011544041.2 | XP_011542343.1 | 7 | 6780 | |||
XM_011544043.2 | XP_011542345.1 | 9 | 7113 | |||
XM_011544044.2 | XP_011542346.1 | 8 | 6884 | |||
XM_011544045.2 | XP_011542347.1 | 8 | 7009 | |||
XM_011544046.2 | XP_011542348.1 | 6 | 6717 | |||
XM_011544047.2 | XP_011542349.1 | 6 | 6653 | |||
XM_017000152.1 | XP_016855641.1 | 8 | 9706 | |||
XM_017000153.1 | XP_016855642.1 | 7 | 6784 | |||
XM_017000154.1 | XP_016855643.1 | 6 | 6478 | |||
XM_017000155.1 | XP_016855644.1 | 7 | 6821 | |||
XM_017000156.1 | XP_016855645.1 | 7 | 6785 | |||
XM_017000157.1 | XP_016855646.1 | 5 | 6386 | |||
XM_017000158.1 | XP_016855647.1 | 5 | 6618 | |||
XM_017000159.1 | XP_016855648.1 | 5 | 6590 | |||
XM_017000160.1 | XP_016855649.1 | 4 | 7440 | |||
XM_017000161.1 | XP_016855650.1 | 6 | 6722 | |||
XM_017000162.1 | XP_016855651.1 | 5 | 6554 | |||
XM_017000163.1 | XP_016855652.1 | 4 | 10416 | |||
GenBank mRNA | AK095715.1 | - | 1 | 1386 |