Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 12 | 6154 | ||
XM_011538078.2 | XP_011536380.1 | 13 | 6156 | |||
XM_017019083.1 | XP_016874572.1 | 12 | 6024 | |||
XM_017019084.1 | XP_016874573.1 | 14 | 5815 | |||
XM_017019085.1 | XP_016874574.1 | 14 | 6035 | |||
XM_017019086.1 | XP_016874575.1 | 14 | 6040 | |||
XM_017019087.1 | XP_016874576.1 | 12 | 5906 | |||
GenBank mRNA | AL050074.1 | - | 1 | 731 |