Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524664.2 | XP_011522966.1 | 1 | 372 | ||
XM_011524666.1 | XP_011522968.1 | 1 | 160 | |||
XM_011524676.1 | XP_011522978.1 | 1 | 160 | |||
XM_017024508.1 | XP_016879997.1 | 1 | 372 | |||
XM_017024509.1 | XP_016879998.1 | 1 | 372 | |||
XM_017024520.1 | XP_016880009.1 | 1 | 372 |