Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1629 | |||
NR_136193.1 | 4 | 2326 | ||||
NR_136194.1 | 4 | 1796 | ||||
NR_136197.1 | 4 | 1700 | ||||
NR_136198.1 | 3 | 1517 | ||||
NR_136199.1 | 5 | 1839 | ||||
NR_136200.1 | 4 | 2329 | ||||
NR_136201.1 | 4 | 2496 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 2237 | ||
AK021601.1 | - | 1 | 1976 | |||
DQ831955.1 | - | 1 | 389 | |||
DQ831956.1 | - | 1 | 389 | |||
EF529495.1 | - | 4 | 629 | |||
EF529496.1 | - | 4 | 552 | |||
EF529497.1 | - | 4 | 500 | |||
EF529498.1 | - | 4 | 764 | |||
EF529506.1 | - | 4 | 629 | |||
EF529507.1 | - | 4 | 552 | |||
EF529508.1 | - | 4 | 629 | |||
EF529509.1 | - | 4 | 552 | |||
EU000853.1 | - | 4 | 1542 | |||
EU000854.1 | - | 4 | 1542 | |||
EU000855.1 | - | 3 | 2634 | |||
EU000856.1 | - | 3 | 2634 |