Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1757 | |||
NR_136193.1 | 4 | 2454 | ||||
NR_136194.1 | 4 | 1924 | ||||
NR_136197.1 | 4 | 1828 | ||||
NR_136198.1 | 3 | 1645 | ||||
NR_136199.1 | 5 | 1967 | ||||
NR_136200.1 | 4 | 2457 | ||||
NR_136201.1 | 4 | 2624 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 2365 | ||
AK021601.1 | - | 1 | 2104 | |||
DQ831955.1 | - | 1 | 517 | |||
DQ831956.1 | - | 1 | 517 | |||
EF529495.1 | - | 4 | 757 | |||
EF529496.1 | - | 4 | 680 | |||
EF529497.1 | - | 4 | 628 | |||
EF529498.1 | - | 4 | 892 | |||
EF529506.1 | - | 4 | 757 | |||
EF529507.1 | - | 4 | 680 | |||
EF529508.1 | - | 4 | 757 | |||
EF529509.1 | - | 4 | 680 | |||
EU000853.1 | - | 4 | 1670 | |||
EU000854.1 | - | 4 | 1670 | |||
EU000855.1 | - | 3 | 2762 | |||
EU000856.1 | - | 3 | 2762 |