Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1566 | |||
NR_136193.1 | 4 | 2263 | ||||
NR_136194.1 | 4 | 1733 | ||||
NR_136197.1 | 4 | 1637 | ||||
NR_136198.1 | 3 | 1454 | ||||
NR_136199.1 | 5 | 1776 | ||||
NR_136200.1 | 4 | 2266 | ||||
NR_136201.1 | 4 | 2433 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 2174 | ||
AK021601.1 | - | 1 | 1913 | |||
DQ831955.1 | - | 1 | 326 | |||
DQ831956.1 | - | 1 | 326 | |||
EF529495.1 | - | 4 | 566 | |||
EF529496.1 | - | 4 | 489 | |||
EF529497.1 | - | 4 | 437 | |||
EF529498.1 | - | 4 | 701 | |||
EF529506.1 | - | 4 | 566 | |||
EF529507.1 | - | 4 | 489 | |||
EF529508.1 | - | 4 | 566 | |||
EF529509.1 | - | 4 | 489 | |||
EU000853.1 | - | 4 | 1479 | |||
EU000854.1 | - | 4 | 1479 | |||
EU000855.1 | - | 3 | 2571 | |||
EU000856.1 | - | 3 | 2571 |