Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199642.1 | NP_001186571.1 | 21 | 4999 | ||
NM_183357.2 | NP_899200.1 | 21 | 5974 | |||
XM_005247077.3 | XP_005247134.1 | 22 | 6136 | |||
XM_005247078.1 | XP_005247135.1 | 22 | 5074 | |||
XM_006713483.1 | XP_006713546.1 | 22 | 5070 | |||
XM_006713484.1 | XP_006713547.1 | 22 | 5075 | |||
XM_011512359.2 | XP_011510661.1 | 22 | 5934 | |||
XM_011512360.2 | XP_011510662.1 | 22 | 5131 | |||
XM_011512361.1 | XP_011510663.1 | 22 | 5038 | |||
XM_017005638.1 | XP_016861127.1 | 22 | 4966 | |||
XM_017005639.1 | XP_016861128.1 | 22 | 4966 | |||
XM_017005640.1 | XP_016861129.1 | 22 | 5062 | |||
GenBank mRNA | AK098381.1 | - | 1 | 1562 |