Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316312.1 | NP_001303241.1 | 5 | 1509 | ||
GenBank mRNA | BC055419.1 | - | 5 | 1201 |