Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276880.1 | XP_005276937.1 | 1 | 265 | ||
XM_005276881.1 | XP_005276938.1 | 1 | 265 | |||
XM_005276882.1 | XP_005276939.1 | 1 | 265 | |||
XM_005276883.2 | XP_005276940.1 | 1 | 265 | |||
XM_017024739.1 | XP_016880228.1 | 1 | 265 | |||
XM_017024741.1 | XP_016880230.1 | 1 | 265 | |||
GenBank mRNA | X96849.1 | - | 1 | 270 |