Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n322241
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
30
7299
NM_001324418.1
NP_001311347.1
32
7584
NM_001324419.1
NP_001311348.1
32
7581
NM_001324420.1
NP_001311349.1
31
7497
NM_001324421.1
NP_001311350.1
29
7178
NM_016351.5
NP_057435.2
30
7302
NM_021722.5
NP_068368.2
31
7389
NM_021723.4
NP_068369.1
31
7410
XM_005250445.3
XP_005250502.1
30
7258
XM_006716028.3
XP_006716091.1
32
7456
XM_006716029.3
XP_006716092.1
29
7134
XM_011516318.1
XP_011514620.1
33
7587
XM_011516319.2
XP_011514621.1
33
7671
XM_011516320.2
XP_011514622.1
31
7369
XM_011516321.2
XP_011514623.1
31
7345
XM_011516322.1
XP_011514624.1
33
7500
XM_011516323.1
XP_011514625.1
32
7392
XM_011516324.1
XP_011514626.1
33
7259
XM_017012329.1
XP_016867818.1
33
7443
XM_017012330.1
XP_016867819.1
32
7440
XM_017012331.1
XP_016867820.1
32
7473
XM_017012332.1
XP_016867821.1
32
7389
XM_017012333.1
XP_016867822.1
30
7178
XM_017012334.1
XP_016867823.1
32
7356
XM_017012335.1
XP_016867824.1
32
7332
XM_017012336.1
XP_016867825.1
31
7305
XM_017012337.1
XP_016867826.1
30
7121
XM_017012338.1
XP_016867827.1
31
7381
XM_017012339.1
XP_016867828.1
31
7245
XM_017012340.1
XP_016867829.1
22
6318
XM_017012341.1
XP_016867830.1
29
6922
GenBank mRNA
AL133090.1
-
1
1064
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline