Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006609.4 | NP_006600.3 | 1 | 29 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC017814.1 | - | 1 | 271 | Chr. 2 - 127343065 - 127343472 [-] on Build GRCh38 |