Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 1 | 119 | |||
GenBank mRNA | AK096279.1 | - | 1 | 102 | ||
BC033386.1 | - | 1 | 102 |