Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000855.2 | NP_000846.1 | 8 | 12120 | ||
NM_001256424.1 | NP_001243353.1 | 9 | 12213 | |||
GenBank mRNA | AK001875.1 | - | 1 | 638 | ||
AK022090.1 | - | 1 | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU135934.1 | - | 1 | 638 | Chr. 11 - 106677892 - 106678655 [-] on Build GRCh38 |