Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000855.2 | NP_000846.1 | 8 | 11772 | ||
NM_001256424.1 | NP_001243353.1 | 9 | 11865 | |||
GenBank mRNA | AK001875.1 | - | 1 | 290 | ||
AK022090.1 | - | 1 | 35 |