Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018676.1 | - | 1 | 20 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF790481.1 | - | 17 | Chr. - - [-] on Build |