Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120582.1 | 3, 4 | 645 | |||
GenBank mRNA | BC038579.2 | - | 3, 4 | 645 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000631195.1 | - | 645 | Chr. - - [-] on Build |