Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293228.1 | NP_001280157.1 | 1 | 77 | ||
NM_001293231.1 | NP_001280160.1 | 1 | 77 | |||
NM_001293233.1 | NP_001280162.1 | 1 | 77 | |||
NM_005378.5 | NP_005369.2 | 1 | 77 | |||
GenBank mRNA | M55330.1 | - | 1 | 77 |