Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319157.1 | NP_001306086.1 | 3 | 791 | ||
GenBank mRNA | AK097281.1 | - | 1 | 502 | ||
AK294214.1 | - | 3 | 749 |