Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272111.4 | XP_005272168.1 | 8 | 5524 | ||
XM_017009962.1 | XP_016865451.1 | 7 | 5390 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK022140.1 | - | 1 | 2168 | Chr. 5 - 93860669 - 93863825 [-] on Build GRCh38 |