Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324046.1 | NP_001310975.1 | 16 | 11395 | ||
NM_001324047.1 | NP_001310976.1 | 15 | 11335 | |||
NM_001324048.1 | NP_001310977.1 | 16 | 11421 | |||
NM_001324049.1 | NP_001310978.1 | 17 | 11544 | |||
NM_004671.4 | NP_004662.2 | 14 | 11223 | |||
NR_136684.1 | 13 | 11138 | ||||
GenBank mRNA | AF086240.1 | - | 1 | 632 |