Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528232.1 | XP_011526534.1 | 1 | 73 | ||
XM_017027189.1 | XP_016882678.1 | 1 | 73 | |||
GenBank mRNA | AK096732.1 | - | 1 | 73 |