Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110058.1 | 5 | 1031 | |||
GenBank mRNA | BC040912.1 | - | 5 | 1031 |